Learn More
MilliporeSigma™ anti-Myelin Basic Protein (MBP), Polyclonal, ™ (Chemicon™)
Supplier: MilliporeSigma™ AB980/DEL
Description
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Myelin Basic Protein (MBP) | |
Unconjugated | |
Human myelin basic protein from brain. | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
500 μL | |
Neuroscience | |
Human, Mouse |