Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Proteolipid Protein Mouse anti-Mammals, Clone: PLPC1, MilliporeSigma™

Catalog No. MAB388100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB388100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB388100UG Supplier MilliporeSigma Supplier No. MAB388100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Myelin Proteolipid Protein using this Anti-Myelin Proteolipid Protein Antibody, C-terminus, clone PLPC1 validated for use in immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Proteolipid Protein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone PLPC1
Conjugate Unconjugated
Formulation Purified mouse immunoglobulin in PBS, pH 7.2 containing 0.09% sodium azide.
Gene Accession No. NM_000533.3
Gene Symbols PLP, PMD, MMP, SPG2, PLP/DM20
Host Species Mouse
Immunogen Myelin proteolipid protein, synthetic peptide CGRGTKF corresponding to the C-terminal peptide.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.