Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloid differentiation-2 (MD-2) Mouse anti-Human, Clone: IIC1, MilliporeSigma™

Catalog No. MABF117MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF117MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF117MI Supplier MilliporeSigma Supplier No. MABF117

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect Myeloid differentiation-2 (MD-2) using this Anti-Myeloid differentiation-2 (MD-2) Antibody, clone IIC1 validated for use in flow cytometry, immunocytochemistry, western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid differentiation-2 (MD-2)
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone IIC1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001182726
Gene Symbols LY96, ESOP1, MD2
Host Species Mouse
Immunogen Mouse MD-2 and HEK293 cells stably expressing TLR4 and MD-2.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.