Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1224MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1224MI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1224MI Supplier MilliporeSigma Supplier No. AB1224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Myeloperoxidase Antibody, human white cell is an antibody against Myeloperoxidase for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloperoxidase
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Monospecific antiserum purified by delipidation and defibrination. Lyophilized from 0.02M Potassium Phosphate buffer, 0.15M Sodium Chloride, pH 7.2, 0.01% Sodium Azide. Reconstitute with 2.0 mL of sterile distilled water.
Gene Symbols MPO; myeloperoxidase
Host Species Rabbit
Immunogen Myeloperoxidase (Human White Cells)
Quantity 2 mL
Primary or Secondary Primary
Gene ID (Entrez) NM_000250.1
Target Species Human
Content And Storage 2-8°C in undiluted aliquots, −20°C for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.