Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase Mouse anti-Human, Unlabeled, Clone: 1B10, BD

Catalog No. BDB556035 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556035 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556035 Supplier BD Biosciences Supplier No. 556035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with myeloperoxidase (MPO), a hemoprotein of 140kDa, composed of two heavy subunits of 52kDa and two light chains of 15kDa. MPO is stored in primary azurophilic granules of neutrophils and plays a major role in the bactericidal activity of neutrophils during phagocytosis. It catalyzes the generation of hypochlorous acid, a powerful oxidant. 1B10 antibody detects MPO in neutrophils, monocytes and HL-60 cells. This reagent is suitable for immunohistochemical staining of acetone-fixed, frozen tissue section and/or formalin-fixed,
paraffin-embedded tissue sections with TUF pretreatment.

Host Species: Mouse
Clone: 1B10
Isotype: IgG1 κ
Species Reactivity: Human

Immunohistochemistry, Immunohistochemistry (Frozen), Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloperoxidase
Applications Immunohistochemistry
Classification Monoclonal
Clone 1B10
Concentration 0.5mg/mL
Conjugate Unconjugated
Description MPO
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.