Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase Mouse anti-Human, FITC, Clone: 5B8, BD

Catalog No. BDB340580 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340580 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340580 Supplier BD Biosciences Supplier No. 340580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-human myeloperoxidase (MPO) reacts with intracellular MPO, a hemoprotein consisting of two molecular subunits, 60kDa and 12kDa MPO is a major enzyme involved in the inflammatory responses of polymorphonuclear leucocytes. It catalyzes the production of hypochlorous acid, a potent microbicidal agent.MPO is a lysosomal enzyme localized in the azurophilic granules of mature granulocytes and is present at low density in circulating monocytes. Detectable MPO is also present in myeloblasts from some cases of acute myeloid leukemia.

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloperoxidase
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 5B8
Concentration 12.5μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Commercially purified myeloperoxidase
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.