Learn More
anti-Myeloperoxidase, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB9535500UL

Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human, Monkey
Immunogen: Myeloperoxidase isolated from human granulocytes
Applications:
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Specifications
Myeloperoxidase | |
2mg/mL | |
Rabbit | |
500 μL | |
Primary | |
Purified |
Polyclonal | |
Unconjugated | |
Myeloperoxidase isolated from human granulocytes | |
Cancer and Tumor Biology, Cardiovascular, Immunology | |
Human, Monkey, Mouse, Rat | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.