Learn More
myosin, light chain 4, alkali; atrial, embryonic, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Myosin is a hexameric ATPase cellular motor protein. It is composed of two myosin heavy chains, two nonphosphorylatable myosin alkali light chains, and two phosphorylatable myosin regulatory light chains. This gene encodes a myosin alkali light chain that is found in embryonic muscle and adult atria. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | myosin, light chain 4, alkali; atrial, embryonic |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human MYL4 protein. |
| Formulation | No additive |
| Gene | MYL4 |
| Gene Accession No. | NM_001002841.1 |
| Gene Alias | ALC1/AMLC/GT1/PRO1957 |
| Gene Symbols | MYL4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.