Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myosin, light chain 9, regulatory, Mouse, Clone: 3F2, Abnova™

Catalog No. 89001177 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-177 Supplier Abnova Corporation Supplier No. H00010398M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MYL9.

Myosin, a structural component of muscle, consists of two heavy chains and four light chains. The protein encoded by this gene is a myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. The encoded protein binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MSSKRAKAKTTKKRPQRATSNVFAMFDQSQIQEFKEAFNMIDQNRDGFIDKEDLHDMLASLGFIHEDHLRELLTTMGDRFTDEEVDEMYREAPIDKKGNFNYVEFTRILKHGAKDKDD

Specifications

Antigen myosin, light chain 9, regulatory
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MYL9.
Formulation PBS with no preservative; pH 7.4
Gene MYL9
Gene Accession No. BC002648
Gene Alias LC20/MGC3505/MLC2/MRLC1/MYRL2
Gene Symbols MYL9
Host Species Mouse
Immunogen MYL9 (AAH02648.1, 1 a.a. ∼ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10398
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.