Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyoD1 Mouse anti-Chicken, Human, Mouse, Rat, Clone: 5.2F, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3878
Description
Anti-MyoD1 Antibody is a high quality Mouse Monoclonal Antibody for the detection of MyoD1 and has been validated in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
Specifications
MyoD1 | |
Monoclonal | |
Unconjugated | |
NM_002478.4 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
5.2F | |
Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. | |
MYOD1, MYOD, PUM, Myf-3, MYF3 | |
Recombinant protein from mouse MyoD1. | |
100 μg | |
Chicken, Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction