Learn More
MYOD1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MYOD1 peptide using ARM Technology.
Supplier: Abnova Corporation H00004654K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a nuclear protein that belongs to the basic helix-loop-helix family of transcription factors and the myogenic factors subfamily. It regulates muscle cell differentiation by inducing cell cycle arrest, a prerequisite for myogenic initiation. The protein is also involved in muscle regeneration. It activates its own transcription which may stabilize commitment to myogenesis. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
MYOD1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
MYOD1 | |
MYOD1 | |
A synthetic peptide of human MYOD1 | |
RUO | |
4654 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
myogenic differentiation 1 | |
MYOD1 | |
MYF3, MYOD, PUM, bHLHc1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.