Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin 2 Mouse anti-Human, Mouse, Rat, Clone: 5B11.1, MilliporeSigma™

Catalog No. MABT25MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT25MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT25MI Supplier MilliporeSigma Supplier No. MABT25
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Myosin 2 Antibody, clone 5B11.1 is validated for use in Western Blotting, immunohistochemistry for the detection of Myosin 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin 2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5B11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_060004
Gene Symbols MYH2, MYHSA2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to mouse Myosin 2.
Purification Method Purified by ion-exchange chromatography.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.