Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Mouse anti-Human, Clone: 5B9 (aka 2C8), MilliporeSigma™

Catalog No. MAB1548MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1548MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1548MI Supplier MilliporeSigma Supplier No. MAB1548
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Myosin using this Anti-Myosin Antibody, heavy chain β validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5B9 (aka 2C8)
Conjugate Unconjugated
Formulation Supernatant in RPMI 1640, 10% FCS, 0.01% sodium azide.
Gene Accession No. NM_000257.2
Gene Symbols MYH7, Myosin-7, MYHCB, MyHC-beta, DKFZp451F047, CMH1, CMD1S, MGC138378, MPD1, MGC138376
Host Species Mouse
Immunogen Human ventricular myosin
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.