Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Mouse anti-Human, Rat, Feline, Clone: NOQ7.5.4D, MilliporeSigma™

Catalog No. MAB1628MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1628MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1628MI Supplier MilliporeSigma Supplier No. MAB1628
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Myosin Antibody, slow muscle, clone NOQ7.5.4D is an antibody against Myosin for use in RIA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin
Applications Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone NOQ7.5.4D
Conjugate Unconjugated
Formulation Liquid.
Gene Accession No. NM_000257.2
Gene Symbols MYH7, Myosin-7, MYHCB, MyHC-beta, CMD1S, MGC138378, MPD1, DKFZp451F047, MGC138376, CMH1
Host Species Mouse
Immunogen Myosin purified from myofibrils isolated from histochemically mixed human skeletal muscle.
Quantity 100 μg
Primary or Secondary Primary
Target Species Feline, Human, Rat
Content And Storage 2-8°C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.