Learn More
Myosin Light Chain Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Myosin Light Chain |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.02M Potassium phosphate, pH 7.2 with 0.15M Sodium chloride and 0.01% sodium azide. |
| Gene Symbols | MYL6B; MLC1SA; MLC1sa |
| Host Species | Rabbit |
| Immunogen | Synthetic phosphopeptide from human myosin, smooth and non-muscle regulatory light chain. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.