Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin X Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1094MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1094MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1094MI Supplier MilliporeSigma Supplier No. AB1094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Myosin X using this Anti-Myosin X Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin X Antibody
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum containing 0.05% NaN3.
Gene Symbols MYO10
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide to amino acids 936-953 of human Myosin X.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) AAF37875
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.