Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYPT1 Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB612165 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612165 150 μg
BDB612164 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612165 Supplier BD Biosciences Supplier No. 612165
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein phosphatase-1 (PP1) is a major serine/threonine phosphatase that is involved in various cellular processes in eukaryotic cells. The PP1 catalytic subunit may be directed to particular subcellular locations by targeting subunits, which can modify the substrate specificity of the enzyme. The PP1 enzyme associated with the myofibrils of striated and smooth muscle forms a complex with Myosin Phosphatase Targeting subunit 1 (MYPT1). MYPT1 contains seven ankyrin repeats in the N-terminal region, and a leucine zipper (LZ) motif in the C-terminus. Alternative splicing of the MYPT1 mRNA leads to expression of a truncated MYPT1 that lacks the leucine zipper region. The expression of MYPT1 splice variants correlate with a smooth muscle phenotype. In the tonic contracting chicken aorta, only the full length MYPT1 is expressed, while in the phasic contracting chicken gizzard, only the truncated MYPT1 is expressed. The leucine zipper motif may be required for association with cGMP-dependent protein kinase I. Thus, MYPT1 may direct not only PP1 enzymatic activity toward myosin, but also may regulate the cGMP responsiveness of PP1 through expression of alternative splice variants.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPT1
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Myosin Phosphatase Targeting subunit 1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat MYPT1 aa. 723-840
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.