Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myelin transcription factor 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-211 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-211 Supplier Abnova Corporation Supplier No. H00004661A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MYT1.

The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. [provided by RefSeq

Sequence: DYASFDAQVFGKRMLAPKIQTSETSPKAFQCFDYSQDAEAAHMAATAILNLSTRCWEMPENLSTKPQDLPSKSVDIEVDENGTLDLSMHKHRKRENAFPS

Specifications

Antigen myelin transcription factor 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MYT1.
Formulation 50% glycerol
Gene MYT1
Gene Accession No. NM_004535
Gene Alias C20orf36/MTF1/MYTI/PLPB1
Gene Symbols MYT1
Host Species Mouse
Immunogen MYT1 (NP_004526, 586 a.a. ∼ 685 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4661
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.