Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-N-Acetyl-D-Glucosamine Kinase/NAGK, Polyclonal, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB10075497
Description
N-Acetyl-D-Glucosamine Kinase/NAGK Polyclonal specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
N-Acetyl-D-Glucosamine Kinase/NAGK | |
Polyclonal | |
Western Blot | |
Q53HD5 | |
NAGK | |
The immunogen for this antibody was a sequence from 1a.a. to 240a.a. from N Acetylglucosamine kinase(a 344a.a. long protein) | |
0.05 mg | |
metabolism | |
55577 | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
PBS with No Preservative | |
EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910 | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
N acetylglucosamine kinase | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction