Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-N-Ethylmaleimide-Sensitive Factor Clone: NSF-1,

Catalog No. NB21100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NB21100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB21100UG Supplier MilliporeSigma™ Supplier No. NB21100UG

Mouse Monoclonal Antibody

Specifically detects N-Ethylmaleimide-Sensitive Factor Clone: NSF-1 in Human, Rat samples, and it is validated for ELISA, Immunoprecipitation, Immuno Blotting

Synaptic vesicle proteins can be divided into one of two classes; transport proteins involved in neurotransmitter uptake, and trafficking proteins which perform functions in vesicle docking, fusion, and budding. For example, synaptophysin may function in the process of neurotransmitter release, while synaptobrevin (VAMP) is believed to play a role in synaptic vesicle docking and exocytotic membrane fusion. Synaptobrevin has also been shown to bind synaptophysin with high specificity. Synapsins are highly abundant vesicle membrane proteins that are believed to regulate neurotransmitter release through phosphorylation-dependent interactions with cytoskeletal elements. Other proteins involved in vesicle exocytosis include syntaxin 1 and SNAP-25. In conjunction with synaptobrevin, these proteins form the SNARE (SNAP receptor) complex, which recruits other cytosolic factors including SNAP proteins (soluble NSF attachment proteins) and NSF (N-ethylmaleimide-sensitive factor). NSF acts in concert with SNAPs and SNAREs for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicle-to-plasma membrane transport. In its functional form, NSF is a homotrimer of 76kDa subunits. Each subunit contains an N-terminal domain and two homologous ATP binding domains (D1 and D2). Vesicle-to-target membrane docking occurs when the vesicle SNARE binds to its cognate target membrane SNARE. SNAPs α and γ then bind to this docking complex and mediate the binding of NSF and thus the formation of a 20 S fusion particle. It is thought that once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-Ethylmaleimide-Sensitive Factor
Applications ELISA, Immunoblot, Immunoblot, Immunoprecipitation
Classification Monoclonal
Clone NSF-1
Conjugate Unconjugated
Formulation In 150mM NaCl, 20mM sodium phosphate, 50% glycerol, pH 7.5.
Host Species Mouse
Immunogen recombinant, human N-ethylmaleimide-senstive factor
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽76kDa NSF-1 protein in human fibroblasts, MDBK, and PC12 cells.
Target Species Human, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.