Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Myc Mouse anti-Human, Clone: NCM II 100, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE333
Description
Use Anti-N-Myc Antibody, clone NCM II 100 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect N-Myc also known as N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, bHLHe37.
Specifications
N-Myc | |
Monoclonal | |
Unconjugated | |
NP_005369 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
NCM II 100 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
MYCN, BHLHE37, NMYC | |
Recombinant protein corresponding to human N-Myc. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction