Learn More
Description
Specifications
Specifications
| Antigen | N-WASP, phospho-specific [Ser484/Ser485] |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | This antibody was cross-adsorbed to unphosphorylated-N-WASP (S484/S485) peptide (without carrier) then affinity purified using phospho-N-WASP (S484/S485) peptide (without carrier). Antibody is supplied in 100 #956;l phosphate-buffered saline, 50% gl |
| Gene Symbols | WASL; DKFZp779G0847; MGC48327; N-WASP; NWASP |
| Host Species | Rabbit |
| Immunogen | Phospho-N-WASP (S484/S485) synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 484 and 485 of human N-WASP. The human WASP sequence has a similar peptide sequence surrounding serine 483 and 484. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.