Learn More
Description
Specifications
Specifications
| Antigen | N-WASP, phospho-specific [Tyr256] |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | This antibody was cross-adsorbed to phosphotyrosine then affinity purified using phospho-N-WASP (Tyr256) peptide (without carrier). Antibody is supplied in 100 #956;l phosphate-buffered saline, 50% glycerol, 1mg/mL BSA, and 0.05% sodium azide. |
| Gene Symbols | WASL; DKFZp779G0847; MGC48327; N-WASP; NWASP |
| Host Species | Rabbit |
| Immunogen | Phospho-N-WASP (Tyr256) synthetic peptide (coupled to BSA) corresponding to amino acid residues around tyrosine 256 of human N-WASP. The human WASP sequence has a two amino acid difference in the same region surrounding tyrosine 291. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.