Learn More
Description
Specifications
Specifications
| Antigen | N-WASP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity-purified antibody is supplied in 100 #956;l phosphate-buffered saline, 50% glycerol, 1mg/mL BSA, and 0.05% sodium azide. |
| Gene Symbols | WASL; DKFZp779G0847; MGC48327; N-WASP; NWASP |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminus of human N-WASP. This N-WASP peptide sequence is 100% homologous to rat and mouse N-WASP, and has low homology to the corresponding region in the human WASP sequence |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.