Learn More
N-6 adenine-specific DNA methyltransferase 1 (putative), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human N6AMT1 protein.
Supplier: Abnova Corporation H00029104D01P
Description
The protein encoded by this gene belongs to the methyltransferase superfamily. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq
Sequence: MAGENFATPFHGHVGRGAFSDVYEPAEDTFLLLDALEAAAAELAGVEICLEVGSGSGVVSAFLASMIGPQALYMCTDINPEAAACTLETARCNKVHIQPVITDLVGSHGIEAAWAGGRNGREVMDRFFPLVPDLLSPRGLFYLVTIKENNPEEILKIMKTKGLQGTTALSRQAGQETLSVLKFTKSSpecifications
N-6 adenine-specific DNA methyltransferase 1 (putative) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human N6AMT1 protein. | |
N6AMT1 | |
C21orf127/HEMK2/MGC19995/MTQ2/N6AMT/PRED28 | |
Rabbit | |
Affinity Purified | |
RUO | |
29104 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011554.1 | |
N6AMT1 | |
N6AMT1 (AAH11554.1, 1 a.a. ∼ 186 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.