Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Na+/H+ Exchanger-2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3083
Description
Anti-Na+/H+ Exchanger-2 Antibody is an antibody against Na+/H+ Exchanger-2 for use in ELISA and western blotting.
Specifications
Na+/H+ Exchanger-2 | |
Polyclonal | |
Liquid in PBS with 0.1% BSA | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
ELISA, Western Blot | |
Unconjugated | |
SLC9A2; Na; +H; NHE-2; NHE2 | |
A 20 AA peptide within the cytoplasmic, C-terminal domain of the rat NHE2 (Collins et al. 1990; Wang et al. 1993; Ghishan et al. 1995; Chris Yun et al. 1995; Josette & Pouyssegur 1995). | |
50 μg | |
NM_003048.3 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction