Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+/H+ Exchanger-3 Mouse anti-Rat, Rabbit, Clone: 4F5, MilliporeSigma™

Catalog No. MAB3136MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3136MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3136MI Supplier MilliporeSigma Supplier No. MAB3136
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Na+/H+ Exchanger-3 Antibody, isoform NHE3, clone 4F5 detects level of Na+/H+ Exchanger-3 and has been published and validated for use in immunocytochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+/H+ Exchanger-3
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4F5
Conjugate Unconjugated
Formulation Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_001082697.1
Gene Symbols SLC9A3, NHE3, NHE-3, MGC126718, Na, MGC126720, +H
Host Species Mouse
Immunogen Maltose Binding Protein fusion protein containing C-terminal 131 amino acids of rabbit NHE3.
Quantity 100 μg
Primary or Secondary Primary
Target Species Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.