Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+/H+ Exchanger-3 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3085MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3085MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3085MI Supplier MilliporeSigma Supplier No. AB3085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Na+/H+ Exchanger-3 Antibody is validated for use in ELISA, western blotting for the detection of Na+/H+ Exchanger-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+/H+ Exchanger-3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA.
Gene Symbols SLC9A3; Na; NHE3; MGC126720; NHE-3; +H; MGC126718
Host Species Rabbit
Immunogen A 22 AA peptide within the cytoplasmic, c-terminal domain of the rat NHE3 (Brant et al. 1995; Orlowsji et al. 1992; Chris Yun et al. 1995; Josette & Pouyssegur 1995). Control peptide is available, cat# AG779.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_004165
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.