Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Na+/K+ ATPase α-1 Mouse anti-Canine, Human, Mouse, Monkey, Pig, Rat, Rabbit, Xenopus, AlexaFluor™488, Clone: C464.6
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 16243
Description
This Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6, Alexa Fluor 488 is validated for use in flow cytometry, immunocytochemistry, western blotting for the detection of Na+/K+ ATPase α-1.
Specifications
Na+/K+ ATPase α-1 | |
Monoclonal | |
Alexa Fluor 488 | |
ATP1A3, RDP, IIIsubunit)., MGC13276, DYT12, Na, +K | |
Purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla. | |
25 μg | |
Canine, Human, Monkey, Mouse, Pig, Rabbit, Rat, Xenopus species | |
Purified |
Flow Cytometry, Immunocytochemistry, Western Blot | |
C464.6 | |
NM_152296.3 | |
Mouse | |
Protein G purfied | |
Primary | |
2 years at 4°C from date of shipment | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction