Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+/K+ ATPase α-1 Mouse anti-Human, Rat, Rabbit, Mouse, Monkey, Xenopus, Pig, Canine, Sheep, Clone: C464.6, MilliporeSigma™

Catalog No. 05369MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-369-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-369-MI Supplier MilliporeSigma Supplier No. 05369
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Na+/K+ ATPase α-1 (sodium/potassium ATPase subunit alpha 1) using this Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6 validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+/K+ ATPase α-1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C464.6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.02Mphosphate buffer, pH 7.6, 0.25MNaCl, 0.1% sodium azide. Frozen at
−20deg;C.
Produced from a culture supernatant of a hybridoma produced by fusing Ag8 mouse myeloma cells with immunized
Gene Accession No. NM_001001586
Gene Symbols ATP1A3, MGC13276, RDP, EC 3.6.3.9, DYT12
Host Species Mouse
Immunogen Purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla.
Purification Method Protein G chromatography
Quantity 50 μg
Primary or Secondary Primary
Target Species Canine, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.