Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+/K+ ATPase Β-1 Mouse anti-Bovine, Canine, Human, Rat, Rabbit, Clone: C464.8, MilliporeSigma™

Catalog No. 05382MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-382-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-382-MI Supplier MilliporeSigma Supplier No. 05382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Na+/K+ ATPase β-1 Antibody, clone C464.8 is validated for use in immunohistochemistry, western blotting for the detection of Na+/K+ ATPase β-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+/K+ ATPase β-1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone C464.8
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in PBS with 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_001677
Gene Symbols ATP1A3, MGC13276, RDP, IIIsubunit)., DYT12, +K, Na
Host Species Mouse
Immunogen Purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.