Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+/K+ ATPase α-1 Rabbit anti-Bovine, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. 06520MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-520-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-520-MI Supplier MilliporeSigma Supplier No. 06520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Na+/K+ ATPase α-1 Antibody is validated for use in immunocytochemistry, immunoprecipitation, western blotting for the detection of Na+/K+ ATPase α-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+/K+ ATPase α-1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl and 0.05% NaN3.
Gene Symbols ATP1A1; G00-119-711; Na; +K; MGC3285; MGC51750
Host Species Rabbit
Immunogen GST-tagged ATPase.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001001586
Target Species Bovine, Human, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.