Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Na+K+ ATPase α-2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 07674MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-674-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-674-MI Supplier MilliporeSigma Supplier No. 07674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Na+K+ ATPase α-2 Antibody is an antibody against Na+K+ ATPase α-2 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Na+K+ ATPase α-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols ATP1A2; FHM2; MGC59864; KIAA0778; MHP2; Na; +K
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (CKAGQENISVSKRDT) corresponding to amino acids 432-445 of human Na+K+ ATPase α-2, with an N-terminal cysteine added for conjugation purposes.
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000702.2
Target Species Rat
Content And Storage −20°C for up to 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.