Learn More
Na+/K+/Cl- Cotransporter 2 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Na+/K+/Cl− Cotransporter 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA. |
| Gene Symbols | SLC12A1; MGC48843; BSC1; NKCC2 |
| Host Species | Rabbit |
| Immunogen | A 15 amino acid peptide sequence near the C-terminus of rat NKCC2 (Haas et al. 1987; Gamba et al. 1994; Simon et al. 1996; Mount et al. 1995; Igarashi et al. 1995; Mount et al. 1998). The immunogen peptide is available (catalog number AG205) for use in pr |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.