Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAB2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-570 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-570 Supplier Abnova Corporation Supplier No. H00004665B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NAB2 protein.

This gene encodes a member of the family of NGFI-A binding (NAB) proteins, which function in the nucleus to repress transcription induced by some members of the EGR (early growth response) family of transactivators. NAB proteins can homo- or hetero-multimerize with other EGR or NAB proteins through a conserved N-terminal domain, and repress transcription through two partially redundant C-terminal domains. Transcriptional repression by the encoded protein is mediated in part by interactions with the nucleosome remodeling and deactylase (NuRD) complex. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: MHRAPSPTAEQPPGGGDSARRTLQPRLKPSARAMALPRTLGELQLYRVLQRANLLSYYETFIQQGGDDVQQLCEAGEEEFLEIMALVGMATKPLHVRRLQKALREWATNPGLFSQPVPAVPVSSIPLFKISETAGTRKGSMSNGHGSPGEKAGSARSFSPKSPLELGEKLSPLPGGPGAGDPRIWPGRSTPESDVGAGGEEEAGSRWDWGWSRPTGARDGTPACLGRVWMDICRLWGHVQG

Specifications

Antigen NAB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NAB2 protein.
Formulation PBS with no preservative; pH 7.4
Gene NAB2
Gene Accession No. BC007756.2
Gene Alias MADER/MGC75085
Gene Symbols NAB2
Host Species Mouse
Immunogen NAB2 (AAH07756.1, 1 a.a. ∼ 241 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4665
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.