Learn More
nascent polypeptide-associated complex alpha subunit 2, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | nascent polypeptide-associated complex alpha subunit 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant NACAL. |
| Formulation | 50% glycerol |
| Gene | NACA2 |
| Gene Accession No. | NM_199290 |
| Gene Alias | ANAC/MGC71999/NACAL |
| Gene Symbols | NACA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.