Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAGLU, Mouse, Clone: 1B7, Abnova™

Catalog No. 89123237 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-237 Supplier Abnova Corporation Supplier No. H00004669M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NAGLU.

This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate. [provided by RefSeq

Sequence: YQLTLWGPEGNILDYANKQLAGLVANYYTPRWRLFLEALVDSVAQGIPFQQHQFDKNVFQLEQAFVLSKQRYPSQPRGDTVDLAKKIFLKYYPGWVAGS

Specifications

Antigen NAGLU
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NAGLU.
Formulation PBS with no preservative; pH 7.4
Gene NAGLU
Gene Accession No. NM_000263
Gene Alias MPS-IIIB/MPS3B/NAG/UFHSD
Gene Symbols NAGLU
Host Species Mouse
Immunogen NAGLU (NP_000254, 644 a.a. ∼ 742 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4669
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.