Learn More
NAGLU, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NAGLU.
Supplier: Abnova Corporation H00004669A01
Description
This gene encodes an enzyme that degrades heparan sulfate by hydrolysis of terminal N-acetyl-D-glucosamine residues in N-acetyl-alpha-D-glucosaminides. Defects in this gene are the cause of mucopolysaccharidosis type IIIB (MPS-IIIB), also known as Sanfilippo syndrome B. This disease is characterized by the lysosomal accumulation and urinary excretion of heparan sulfate. [provided by RefSeq
Sequence: YQLTLWGPEGNILDYANKQLAGLVANYYTPRWRLFLEALVDSVAQGIPFQQHQFDKNVFQLEQAFVLSKQRYPSQPRGDTVDLAKKIFLKYYPGWVAGSSpecifications
NAGLU | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NAGLU. | |
NAGLU | |
MPS-IIIB/MPS3B/NAG/UFHSD | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000263 | |
NAGLU | |
NAGLU (NP_000254, 644 a.a. ∼ 742 a.a) partial recombinant protein with GST tag. | |
RUO | |
4669 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.