Learn More
N-acetylglutamate synthase, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The N-acetylglutamate synthase gene encodes a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. This gene may regulate ureagenesis by altering NAG availability and, thereby, CPSI activity. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia. [provided by RefSeq
Specifications
Specifications
| Antigen | N-acetylglutamate synthase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant NAGS. |
| Formulation | 50% glycerol |
| Gene | NAGS |
| Gene Accession No. | NM_153006 |
| Gene Alias | AGAS/ARGA/MGC133025 |
| Gene Symbols | NAGS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.