Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5731MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5731MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5731MI Supplier MilliporeSigma Supplier No. AB5731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Nanog using this Anti-Nanog Antibody, N-terminus validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nanog
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal immunoglobulin in buffer containing liquid 0.1% sodium azide.
Gene Symbols FLJ12581; FLJ40451; hNanog
Host Species Rabbit
Immunogen Synthetic peptide from mouse Nanog.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_024865.2
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.