Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanog-Alexa Fluor 488 Rabbit anti-Human, Mouse, Rat, AlexaFluor™488, Polyclonal, MilliporeSigma™

Catalog No. FCABS352A4 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS352A4 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS352A4 Supplier MilliporeSigma Supplier No. FCABS352A4
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Milli-Mark™ Anti-Nanog-Alexa Fluor 488 Antibody, N-terminus is an antibody against Nanog-Alexa Fluor 488 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nanog
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal IgG conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA
Gene Symbols FLJ12581; FLJ40451; hNanog
Host Species Rabbit
Immunogen Linear peptide from mouse Nanog.
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) NP_079141
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.