Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANOG Mouse anti-Human, AlexaFluor™488, Clone: 7F7.1, MilliporeSigma™

Catalog No. MABD24A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD24A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD24A4MI Supplier MilliporeSigma Supplier No. MABD24A4
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NANOG Antibody, clone 7F7.1, Alexa Fluor 488 conjugate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NANOG
Applications Immunocytochemistry
Classification Monoclonal
Clone 7F7.1
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG2a conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/ml BSA.
Gene Accession No. NP_079141
Gene Symbols NANOG
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Nanog
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.