Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP-22 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9304MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9304MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9304MI Supplier MilliporeSigma Supplier No. AB9304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-NAP-22 Antibody, complete sequence is validated for use in western blotting for the detection of NAP-22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP-22
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Semi-purified immunoglobulin. Liquid in TBS containing 0.05% sodium azide.
Gene Symbols BASP1; CAP-23; NAP22; MGC8555; NAP-22; CAP23
Host Species Rabbit
Immunogen Recombinant full length rat NAP-22.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006317.3
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.