Learn More
nucleosome assembly protein 1-like 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NAP1L1 protein.
Supplier: Abnova Corporation H00004673B01
Description
This gene encodes a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation. Alternative splicing of this gene results in several transcript variants; however, not all have been fully described. [provided by RefSeq
Sequence: MADIDNKEQSELDQDLDDVEEVEEEETGEETKLKARQLTVQMMQNPQILAALQERLDGLVETPTGYIESLPRVVKRRVNALKNLQVKCAQIEAKFYEEVHDLERKYAVLYQPLFDKRFEIINAIYEPTEEECEWKPDEEDEISEELKEKAKIEDEKKDEEKEDPKGIPEFWLTVFKNVDLLSDMVQEHDEPILKHLKDIKVKFSDAGQPMSFVLEFHFEPNEYFTNEVLTKTYRMRSEPDDSDPFSFDGPEIMGCTGCQIDWKKGKNVTLKTIKKKQKHKGRGTVRTVTKTVSNDSFFNFFAPPEVPESGDLDDDAEAILAADFEIGHFLRERIIPRSVLYFTGEAIEDDDDDYDEEGEEADEEGEEEGDEENDPDYDPKKDQNPAECKQQSpecifications
nucleosome assembly protein 1-like 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human NAP1L1 protein. | |
NAP1L1 | |
FLJ16112/MGC23410/MGC8688/NAP1/NAP1L/NRP | |
Mouse | |
50 μL | |
Cell Cycle | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC002387 | |
NAP1L1 | |
NAP1L1 (AAH02387, 1 a.a. ∼ 391 a.a) full-length human protein. | |
RUO | |
Yes | |
4673 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.