Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
NARS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514537
Description
Asparagine-tRNA ligase (EC 6.1.1.22) catalyzes the chemical reaction: ATP + L-asparagine + tRNAAsn <---> AMP + diphosphate + L-asparaginyl-tRNAAsn. This enzyme participates in alanine and aspartate metabolism and aminoacyl-tRNA biosynthesis.
            Specifications
| NARS2 | |
| Polyclonal | |
| NARS2 | |
| Probable asparaginyl-tRNA synthetase, mitochondrial [Precursor]; EC 6.1.1.22; Asparagine--tRNA ligase; AsnRS; NARS2 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 79731 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q96I59 | |
| NARS2 | |
| KLH conjugated synthetic peptide between 445-474 amino acids from the C-terminal region of human NARS2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction