Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT1 Mouse, Unlabeled, Clone: 35, BD

Catalog No. BDB610742 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610742 50 μg
BDB610743 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610742 Supplier BD Biosciences Supplier No. 610742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF-4 proteins are required for recognition of mRNA and acceleration of protein translation. This group of proteins consists of the RNA helicase eIF-4A, the RNA-binding protein eIF-4B, the cap-binding protein eIF-4E, and eIF-4G (p220). NAT1 (Novel APOBEC-1 Target no. 1), also known as DAP-5 (Death Associated Protein -5), is homologous to eIF-4G. Amino acid sequence comparison of NAT1 and eIF-4 shows that NAT1 lacks an eIF-4G N-terminal region. This region mediates eIF-4G association with eIF-4E. The highest degree of homology is
within the central portions of NAT1 and eIF-4G, while the lowest degree of homology occurs at the C-terminus. NAT1 more closely resembles a cleaved form of eIF-4G that is involved in cap-independent translation. It is thought that NAT1 is involved in the repression of translation via its inhibition of both cap-dependent and cap-independent translation.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Description Novel APOBEC-1 Target no. 1; DAP-5; Death Associated Protein -5
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NAT1 aa. 672-830
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.