Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-acetyltransferase 6 (GCN5-related) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human NAT6 protein.
Supplier: Abnova Corporation H00024142B01
Description
Sequence: MELILSTSPAELTLDPACQPKLPLDSTCQPEMTFNPGPTGLTLDPEHQPEETPAPSLAELTLEPVHRRPELLDACADLINDQWPRSRTSRLHSLGQSSDAFPLCLMLLSPHPTLEAAPVVVGHARLSRVLNQPQSLLVETVVVARALRGRGFGRRLMEGLEVFARARGFRKLHLTTHDQVHFYTHLGYQLGEPVQGLVFTSRRLPATLLNAFPTAPSPRPPRKAPNLTAQAAPRGPKGPPLPPPPPLPECLTISPPVPSGPPPKSLLETQYQNVRGRPIFWMEKDISpecifications
N-acetyltransferase 6 (GCN5-related) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human NAT6 protein. | |
NAT6 | |
FUS-2/FUS2 | |
Mouse | |
50 μL | |
Yes | |
24142 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC004483 | |
NAT6 | |
NAT6 (AAH04483, 23 a.a. ∼ 308 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction