Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAV2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human NAV2 peptide using ARM Technology.
Supplier: Abnova Corporation H00089797K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene.[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
NAV2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NAV2 | |
NAV2 | |
A synthetic peptide of human NAV2 | |
RUO | |
89797 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
neuron navigator 2 | |
NAV2 | |
FLJ10633, FLJ11030, FLJ23707, FLJ77876, HELAD1, KIAA1419, POMFIL2, RAINB1, STEERIN2, UNC53H2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction