Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

neighbor of BRCA1 gene 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016838 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-838 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-838 Supplier Abnova Corporation Supplier No. H00004077B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NBR1 protein.

The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene. [provided by RefSeq

Sequence: MEPQVTLNVTFKNEIQSFLVSDPENTTWADIEAMVKVSFDLNTIQIKYLDEENEEVSINSQGEYEEALKMAVKQGNQLQMQVHEGHHVVDEAPPPVVGAKRLAARAGKKPLAHYSSLVRVLGSDMKTPEDPAVQSFPLVPCDTDQPQDKPPDWFTSYLETFREQVVNETVEKLEQKLHEKLVLQNPSLGSCPSEVSMPTSEETLFLPENQFSWHIACNNCQRRIVGVRYQCSLCPSYNICEDCEAGPYGHDTNHVLLKLRRPVVGSSEPFCHSKYSTPRLPAALEQVRLQKQVDKNFLKAEKQRLRAEKKQRKAEVKELKKQLKLHRKIHLWNSIHGLQSPKSPLGRPESLLQSNTLMLPLQPCTSVMPMLSAAFVDENLPDGTHLQPGTKFIKHWRMKNTGNVKWSADTKLKFMWGNLTLASTEKKDVLVPCLKAGHVGVVSVEFIAPALEGTYTSHWRLSHKGQQFGPRVWCSIIVDPFPSEESPDNIEKGMISSSKTDDLTCQQEETFLLAKEERQLGEVTEQTEGTAACIPQKAKNVASERELYIPSVDLLTAQDLLSFELLDINIVQELERVPHNTPVDVTPCMSPLPHDSPLIEKPGLGQIEEENEGAGFKALPDSMVSVKRKAENIASVEEAEEDLSGTQFVCETVIRSLTLDAAPDHNPPCRQKSLQMTFALPEGPLGNEKEEIIHIAEEEAVMEEEEDEEDEEEEDELKDEVQSQSSASSEDYIIILPECFDTSRPLGDSMYSSALSQPGLERGAEGKPGVEAGQEPAEAGERLPGGENQPQEHSISDILTTSQTLETVPLIPEVVELPPSLPRSSPCVHHHGSPGVDLPVTIPEVSSVPDQIRGEPRGSSGLVNSRQKSYDHSRHHHGSSIAGGLVKGALSVAASAYKALFAGPPVTAQPIISEDQTAALMAHLFEMGFCDRQLNLRLLKKHNYNILQVVTELLQLNNNDWYSQRY

Specifications

Antigen neighbor of BRCA1 gene 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NBR1 protein.
Formulation PBS with no preservative; pH 7.4
Gene NBR1
Gene Accession No. NM_005899.3
Gene Alias 1A1-3B/KIAA0049/M17S2/MIG19
Gene Symbols NBR1
Host Species Mouse
Immunogen NBR1 (NP_005890.2, 1 a.a. ∼ 966 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4077
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.