Learn More
NBS1 Mouse, Unlabeled, Clone: 34, BD
Mouse Monoclonal Antibody
Specifications
Antigen | NBS1 |
---|---|
Clone | 34 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB611870
|
BD Biosciences
611870 |
50 μg | N/A | N/A | N/A | ||||
Description
Nijmegen Breakage Syndrome (NBS) is characterized by extreme radiation sensitivity and chromosomal instability. The NBS1 gene product, p95/nibrin/NBS1, forms a complex with Rad50 and Mre11. Cells deficient in this complex have problems with DNA double-strand break repair, cell cycle checkpoint control, and telomere length maintenance. NBS1 contains a forkhead-associated domain (FHA) and a breast cancer carboxy-terminal domain (BRCT) in the N-terminal region. Both of these domains have been found in DNA-damage responsive cell cycle checkpoint proteins. The complex containing NBS1, Rad50, and Mre11 possesses manganese-dependent single stranded DNA endonuclease and 3′ to 5′ exonuclease activities. In addition, NBS1 is required for DNA-damage dependent phosphorylation of Mre11. This phosphorylation may be required for proper nuclear localization of the NBS1-Rad50-Mre11 complex to sites of DNA double-strand breaks. NBS1 interacts directly with telomere repeat binding factor, TRF1, via its C-terminal region, and both NBS1 and Mre11 co-localize with TRF1 at promyelocytic leukemia nuclear bodies. Thus, the NBS1-Rad50-Mre11 complex may be important for both DNA damage repair, and telomere length maintenance.
Host Species: Mouse
Clone: 34
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human NBS1 aa. 620-732
Immunofluorescence, Western Blotting
Specifications
NBS1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Nijmegen Breakage Syndrome-1 |
34 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Human NBS1 aa. 620-732 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.